VWR®, Flow Cup viscosity standards
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
99432.260EA 89.8 EUR
99432.260 99437.260 99431.260 99429.260 99430.260 99436.260 99433.260 99434.260 99435.260
VWR®, Flow Cup viscosity standards
Standaarden Standaarden voor viscometrie
Flow Cup viscosity standards are specifically formulated for use with viscosity flow cups such as Ford, Shell and Zahn.
  • Certified in strict accordance with ASTM D2162
  • Density g/ml in accordance with ASTM D1480
  • Density, dynamic viscosity and kinematic viscosity quoted at all temperatures
  • Ensures full compliance to ASTM and IP test method protocols
  • Full traceability to national standards
  • Tamper-evident security packaging
Certified data for kinematic viscosity, dynamic viscosity and density at 20 and 25 °C including relevant flow cup information.
Certificaten: Each standard is dual-certified to ISO 17025 and ISO 17034 under UKAS.
Opgelet: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.