VWR®, Medical Grade viscosity standards
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
99505.180EA 49.1 EUR
99505.180 99505.260 99506.180 99507.180 99508.180 99509.180 99510.180 99511.180 99511.260
VWR®, Medical Grade viscosity standards
Standaarden Standaarden voor viscometrie
Medical Grade viscosity standards provide certified dynamic viscosity data at 25 and 37 °C, are dual-certified to ISO 17025 and ISO 17034 under UKAS and are ideal for the calibration and verification of viscosity measuring equipment.
Full traceability to national standards
Contains an antibacterial agent
Tamper-evident security packaging
Certificaten: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
Opgelet: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.