VWR®, Silicone Rotational viscosity standards
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
99512.271EA 155 EUR
99512.271 99513.271 99514.271 99515.271 99516.271 99517.271 99518.271 99519.271 99520.271 99521.271 99522.271 99523.271 99524.271 99525.271 99526.271
VWR®, Silicone Rotational viscosity standards
Standaarden Standaarden voor viscometrie
Silicone Rotational viscosity standards are specifically formulated for use with equipment with minimal temperature control. Supplied in 600 ml 'ready to use' packs specially designed to eliminate cleaning of laboratory glassware, which is inherently difficult due to the nature of silicone.
  • Ensures full compliance to international test method protocols
  • Full traceability to national standards
  • Tamper-evident security packaging
  • Safer working, less mess, less waste and higher throughput
This range of standards provides both calibration and verification options for rotational viscometer test equipment.
Certificaten: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
Leveringsinformatie: 600 ml of product supplied in a ready to use 'test in pack' container.
Opgelet: Applicable to all spindles with the exception of the L1 spindle, further provision should be made when using the L1 spindle.
The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.