VWR®, Fuel testing standards - gasoline
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
99129.290EA
837
EUR
99129.290
99130.290
VWR®, Fuel testing standards - gasoline
Standaarden
Standaarden voor de petroleumindustrie
Fuel testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Octane Number.
- Certified in accordance with ASTM / EN or IP test method protocols
- Certified by Round Robin method using independent test laboratories
- Ensures good inter-laboratory correlation
- Supplied in tamper-evident, UN compliant security pack