Transport medium, Mediatech (Corning)
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25-500-CI 25-500-CV
MDTC25-500-CIEA 148.87 EUR
MDTC25-500-CI MDTC25-500-CV
Transport medium, Mediatech (Corning)
Media voor microbiologie Geprepareerde media voor microbiologie
Corning® transport medium is formulated using the final ingredient concentrations outlined in CDC's SOP#: DSR-052-01 'Preparation of viral transport medium'.
  • cGMP grade
  • Aseptically sterile-filtered
  • 10⁻³ SAL, labelled 'Sterile A'
Transport medium is an isotonic solution at neutral pH containing protective proteins, antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi.
Order Now
Specificaties
Environmentally Preferable
Storage conditions 2...8 °C