Telkamer, Thoma
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
40440703 40440702 40440701 92044002702 92044001702
631-0927EA 64.3 EUR
631-0927 HECH40440702 HECH40440701 HECH92044002702 HECH92044001702
Telkamer, Thoma
Object- en dekglaasjes Telkamers
These cell counters are designed for blood tests and urine analysis.
  • Four hundred squares of 0,05×0,05 mm (0,0025 m²)
  • Area of central square: 1 mm²
  • Depth of chamber: 0,1 mm (other depths on request)
Bestelinformatie: Other depths are available on request.