VWR® ARISTAR®, ICP Quality Control Standard #1
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456422WEA 579 EUR
456422W 456424B
VWR® ARISTAR®, ICP Quality Control Standard #1
Standaarden Kationische standaarden Standaarden met meerdere elementen
23 Elements 5% nitric acid solution with trace of HF.
23 Elements 100 mg/L : As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, P, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis, tested in ISO Guide 34 /ISO17025 accredited laboratory.