VWR®, ICP multi-element quality control standard 3 in dilute nitric acid ARISTAR® for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456444FEA 1360 EUR
456444F 456442D
VWR®, ICP multi-element quality control standard 3 in dilute nitric acid ARISTAR® for ICP
Standaarden Kationische standaarden Standaarden met meerdere elementen
In 5% nitric acid solution.
15 Elements 100 mg/L : Al, Ba, Cd, Ca, Cr, Co, Cu, Fe, Pb, Mg, Mn, Ni, Na, Ti, Zn
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34 / ISO 17025 accredited facilities.
Supplied with certificate of analysis.