Pumice stone, granules analytical reagent, washed
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26398.293EA 61.4 EUR
26398.293 26398.362
Pumice stone, granules analytical reagent, washed
Pumice stone
Smeltpunt: > 500 °C
MDL Nummer: MFCD00148147
CAS nummer: 1332-09-8
Order Now
Specification Test Results
Heavy metals (as Pb) Max. 50 ppm
Solubility in water Max. 0.5 %
Soluble chlorides Max. 0.05 %