VWR® AVS TITRINORM, Iron Standard Solution, 1000 mg/L Fe in 2% Hydrochloric Acid AVS TITRINORM Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86677.260EA 36.3 EUR
86677.260 86677.180
VWR® AVS TITRINORM, Iron Standard Solution, 1000 mg/L Fe in 2% Hydrochloric Acid AVS TITRINORM Standard for AAS
Standaarden Kationische standaarden IJzer standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%