VWR® AVS TITRINORM, Tin Standard Solution, 1000 mg/L Sn in 20% Hydrochloric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86712.260EA 47.1 EUR
86712.260 86712.180
VWR® AVS TITRINORM, Tin Standard Solution, 1000 mg/L Sn in 20% Hydrochloric Acid, Standard for AAS
Standaarden Kationische standaarden Tin standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%
Order Now
Specificaties
Environmentally Preferable