VWR® AVS TITRINORM, Zinc Standard Solution, 1000 mg/L Zn in dil. Nitric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86725.260EA
47.1
EUR
86725.260
86725.180
VWR® AVS TITRINORM, Zinc Standard Solution, 1000 mg/L Zn in dil. Nitric Acid, Standard for AAS
Standaarden
Kationische standaarden
Zink standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%