G418 disulphate
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
49418.02
49418.03
49418.04
SERA49418.02EA
0
EUR
SERA49418.02
SERA49418.03
SERA49418.04
G418 disulphate
G418 disulphate
Formule:
C₂₀H₄₄N₄O₁₈S₂ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00058314 CAS nummer: 108321-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...