Hydriodinezuur 57% aqueous solution Stabilised analysis_for, distilled
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Hydrogeenjodide
Stabilisatie: 0.75 % HP(OH)2
198370500 198372500 198370010
ACRO198370500EA 40.2 EUR
ACRO198370500 ACRO198372500 ACRO198370010
Hydriodinezuur 57% aqueous solution Stabilised analysis_for, distilled
Hydriodic acid
Formule: HI
MW: 127.91 g/mol
Dichtheid: 1.7 g/cm³
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011347
CAS nummer: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear light yellow to amber Solution (May darken on storage)
Stabilizer 0.75 % H3PO2
Titration with NaOH ≥55.0 %