Hydriodinezuur 57% (w/w) in aqueous solution stabilised
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Stabilisatie:
stab. with hypophosphorous acid 1,5%
CAS No.: 10034-85-2
Molecular Formula: HI
Formula Weight: 127.91
Physical Form: Liquid
Density: 1.701
Sensitivity: Light Sensitive
Appearance: Colourless to yellow
Odour: Pungent
Boiling Point: 126-127°
Merck Reference: 14,4776
UN No.: 1787
MDL No.: MFCD00011347
Formule:
HI Kookpunt: 126…127 °C Dichtheid: 1,701 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011347 CAS nummer: 10034-85-2 EINECS: 233-109-9 UN: 1787 ADR: 8,II Merck Index: 14,4776 |