Rifampicin
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP
A2220.0001 A2220.0005
A2220.0001EA 78.2 EUR
A2220.0001 A2220.0005
Rifampicin
Rifampicin
Formule: C₄₃H₅₈N₄O₁₂
Smeltpunt: 183…188 °C
MDL Nummer: MFCD00151389
CAS nummer: 13292-46-1
EINECS: 236-312-0
Merck Index: 12,08382