Titanium(III) chloride 15% in hydrochloric acid 10%, AnalaR NORMAPUR® analytical reagent
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Titanium trichloride
28660.236EA 58.6 EUR
28660.236 28660.293
Titanium(III) chloride 15% in hydrochloric acid 10%, AnalaR NORMAPUR® analytical reagent
Titanium(III) chloride
Formule: Cl₃Ti
MW: 154.24 g/mol
Kookpunt: 960
Smeltpunt: 440
Dichtheid: 1.2 g/cm³ (20 °C)
MDL Nummer: MFCD00011266
CAS nummer: 7705-07-9
EINECS: 231-728-9
UN: 1789
ADR: 8 II
Merck Index: 12,09620
Order Now
Specification Test Results
Assay 12,5 to 17,5%