VWR® ARISTAR®, Bromide Standard Solution, 1000 mg/L Br- in Water (from NaBr) Standard for Ion Chromatography
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
458042WEA 124 EUR
458042W 458044B
VWR® ARISTAR®, Bromide Standard Solution, 1000 mg/L Br- in Water (from NaBr) Standard for Ion Chromatography
Standaarden Anionische standaarden Bromide standaarden
Br in H₂O.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.