VWR® ARISTAR®, Chloride Standard Solution, 1000 mg/L Cl- in Water (from NaCl) Standard for Ion Chromatography
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
458014SEA 198 EUR
458014S 458012Q
VWR® ARISTAR®, Chloride Standard Solution, 1000 mg/L Cl- in Water (from NaCl) Standard for Ion Chromatography
Standaarden Anionische standaarden Chloride standaarden
(Cl in H₂O).
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.