VWR® ARISTAR®, Nitrite Standard Solution, 1000 mg/L NO₂- in Water (from NaNO₂) Standard for Ion Chromatography
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
458024UEA 280 EUR
458024U 458022S
VWR® ARISTAR®, Nitrite Standard Solution, 1000 mg/L NO₂- in Water (from NaNO₂) Standard for Ion Chromatography
Standaarden Anionische standaarden Nitriet standaarden
(NO₂ in H₂O).
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.