VWR® ARISTAR®, Nitrate Standard Solution, 1000 mg/L NO₃ in Water (from NaNO₃) Standard for Ion Chromatography
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
458034WEA 256 EUR
458034W 458032U
VWR® ARISTAR®, Nitrate Standard Solution, 1000 mg/L NO₃ in Water (from NaNO₃) Standard for Ion Chromatography
Standaarden Anionische standaarden Nitraat standaarden
NO₃ in H₂O.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.