VWR®, Phosphate standard solutions, PO₄³⁻ in Water (from KH₂PO₄) standard for ion chromatography, ARISTAR®
Leverancier: VWR ChemicalsTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
PO₄ in H₂O.
Supplied with certificate of analysis.
Certificaten: Traceable to SRM from NIST, tested in an ISO 17034 / ISO17025 accredited laboratory.