VWR®, Aluminium standard solution, 1000 mg/L Al in 2% hydrochloric acid AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86660.260EA 27.02 EUR
86660.260 86660.180
VWR®, Aluminium standard solution, 1000 mg/L Al in 2% hydrochloric acid AVS TITRINORM standard for AAS
Standaarden Kationische standaarden Aluminium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ISO 17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%