VWR® ARISTAR®, Antimony Standard Solutions for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455022GEA
140
EUR
455022G
455024Y
455034K
VWR® ARISTAR®, Antimony Standard Solutions for ICP
Standaarden
Kationische standaarden
Antimonium standaarden
Sb in HNO₃ 2% tr. tartaric acid.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
Certificaten: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformatie: Supplied with certificate of analysis.