VWR® ARISTAR®, Boron Standard Solution, 10000 mg/L B in Ammonium Hydroxide Solution (max. 1%) (from B(OH₃)), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455132LEA 192 EUR
455132L 455134N
VWR® ARISTAR®, Boron Standard Solution, 10000 mg/L B in Ammonium Hydroxide Solution (max. 1%) (from B(OH₃)), Standard for ICP
Standaarden Kationische standaarden Borium standaarden
H₃BO₃ in H₂O tr. NH₄OH.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.