VWR® AVS TITRINORM, Calcium Standard Solution, 1000 mg/L Ca in dil. Nitric Acid (from Ca(NO₃)₂), Standard for AAS
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR® AVS TITRINORM
86667.260EA 28.1 EUR
86667.260 86667.180
VWR® AVS TITRINORM, Calcium Standard Solution, 1000 mg/L Ca in dil. Nitric Acid (from Ca(NO₃)₂), Standard for AAS
Standaarden Kationische standaarden Calcium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022