VWR® ARISTAR®, Calcium Standard Solution, 1,000 mg/L Ca in dil. Nitric Acid (from CaCO₃), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455162REA 97.2 EUR
455162R 455164T
VWR® ARISTAR®, Calcium Standard Solution, 1,000 mg/L Ca in dil. Nitric Acid (from CaCO₃), Standard for ICP
Standaarden Kationische standaarden Calcium standaarden
CaCO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with Certificate of Analysis.