VWR®, Calcium standard solution, 10000 mg/L Ca in dil. nitric acid (from CaCO₃) ARISTAR® standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455172TEA
217
EUR
455172T
455174V
VWR®, Calcium standard solution, 10000 mg/L Ca in dil. nitric acid (from CaCO₃) ARISTAR® standard for ICP
Standaarden
Kationische standaarden
Calcium standaarden
CaCO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.