VWR® ARISTAR®, Cerium Standard Solution, 1000 mg/L Ce in dil. Nitric Acid (from (NH₄)₂Ce(NO₃)₆), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455182VEA 141.6 EUR
455182V 455184A
VWR® ARISTAR®, Cerium Standard Solution, 1000 mg/L Ce in dil. Nitric Acid (from (NH₄)₂Ce(NO₃)₆), Standard for ICP
Standaarden Kationische standaarden Cerium standaarden
(NH₄)₂ Ce(NO₃)₆ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.