VWR®, Iron standard solution, 10000 mg/L Fe in dil. nitric acid (from Fe), standard for ICP
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR®
455534CEA 597 EUR
455534C 455532A
VWR®, Iron standard solution, 10000 mg/L Fe in dil. nitric acid (from Fe), standard for ICP
Standaarden Kationische standaarden IJzer standaarden
Fe in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022