VWR® ARISTAR®, Europium Standard Solution, 10000 mg/L Eu in dilute Nitric Acid (from Eu2O3), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455352VEA
429
EUR
455352V
455354A
VWR® ARISTAR®, Europium Standard Solution, 10000 mg/L Eu in dilute Nitric Acid (from Eu2O3), Standard for ICP
Standaarden
Kationische standaarden
Europium standaarden
Eu₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.