VWR® ARISTAR®, Gold Standard Solution, 10000 mg/L Au in 10% Hydrochloric Acid (from Au), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455432UEA
0
EUR
455432U
455434W
VWR® ARISTAR®, Gold Standard Solution, 10000 mg/L Au in 10% Hydrochloric Acid (from Au), Standard for ICP
Standaarden
Kationische standaarden
Goud standaarden
Au in HCl 10%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.