VWR® ARISTAR®, Hafnium Standard Solution, 1000 mg/L Hf in Dilute Nitric Acid with hydrofluoric Acid (max. 1%) (from HfO2), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455442WEA 295 EUR
455442W 455444B
VWR® ARISTAR®, Hafnium Standard Solution, 1000 mg/L Hf in Dilute Nitric Acid with hydrofluoric Acid (max. 1%) (from HfO2), Standard for ICP
Standaarden Kationische standaarden Hafnium standaarden
HfO₂ in HNO₃ (2 to 5%) with traces HF.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Order Now
Specificaties
Environmentally Preferable