VWR® ARISTAR®, Copper Standard Solution, 1000 mg/L Cu in dil. Nitric Acid (from Cu), Standard for ICP
Leverancier: VWR ChemicalsTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Cu in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.