VWR® AVS TITRINORM, Copper Standard Solution, 1000 mg/L Cu in dil. Nitric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86673.260EA
32.27
EUR
86673.260
86673.180
VWR® AVS TITRINORM, Copper Standard Solution, 1000 mg/L Cu in dil. Nitric Acid, Standard for AAS
Standaarden
Kationische standaarden
Koper standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%