VWR® ARISTAR®, Copper Standard Solution, 10000 mg/L Cu in dil. Nitric Acid (from Cu), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455294FEA 636 EUR
455294F 455292D
VWR® ARISTAR®, Copper Standard Solution, 10000 mg/L Cu in dil. Nitric Acid (from Cu), Standard for ICP
Standaarden Kationische standaarden Koper standaarden
Cu in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.