VWR® ARISTAR®, Lanthanum Standard Solution, 1000 mg/L La in dil. Nitric Acid (from La₂O₃), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455542CEA 126.6 EUR
455542C 455544E
VWR® ARISTAR®, Lanthanum Standard Solution, 1000 mg/L La in dil. Nitric Acid (from La₂O₃), Standard for ICP
Standaarden Kationische standaarden Lanthanium standaarden
La₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.