VWR® ARISTAR®, Lutetium Standard Solution, 1000 mg/L Lu in dil. Nitric Acid (from Lu₂O₃), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455602UEA 344.4 EUR
455602U 455604W
VWR® ARISTAR®, Lutetium Standard Solution, 1000 mg/L Lu in dil. Nitric Acid (from Lu₂O₃), Standard for ICP
Standaarden Kationische standaarden Lutetium standaarden
Lu₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.