VWR®, Lutetium standard solution, 10000 mg/L Lu in dil. nitric acid (from Lu₂O₃) ARISTAR® standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455612WEA 896 EUR
455612W 455614B
VWR®, Lutetium standard solution, 10000 mg/L Lu in dil. nitric acid (from Lu₂O₃) ARISTAR® standard for ICP
Standaarden Kationische standaarden Lutetium standaarden
Lu₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.