VWR® AVS TITRINORM, Molybdenum Standard Solution, 1000 mg/L Mo in Water, Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86692.260EA 32.97 EUR
86692.260 86692.180
VWR® AVS TITRINORM, Molybdenum Standard Solution, 1000 mg/L Mo in Water, Standard for AAS
Standaarden Kationische standaarden Molybdeen standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%