VWR® AVS TITRINORM, Sodium Standard Solution, 1000 mg/L Na in dil. Nitric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86693.260EA 25.27 EUR
86693.260 86693.180
VWR® AVS TITRINORM, Sodium Standard Solution, 1000 mg/L Na in dil. Nitric Acid, Standard for AAS
Standaarden Kationische standaarden Natrium standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%