VWR® ARISTAR®, Sodium Standard Solutions for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456042XEA
94.8
EUR
456042X
456052Q
456054S
456044Q
VWR® ARISTAR®, Sodium Standard Solutions for ICP
Standaarden
Kationische standaarden
Natrium standaarden
NaNO₃ in HNO₃ 2 to 5%.
Certificaten: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformatie: Supplied with certificate of analysis.