VWR® ARISTAR®, Nickel Standard Solution, 1000 mg/L Ni in dil. Nitric Acid (from Ni), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455722EEA 160 EUR
455722E 455724G
VWR® ARISTAR®, Nickel Standard Solution, 1000 mg/L Ni in dil. Nitric Acid (from Ni), Standard for ICP
Standaarden Kationische standaarden Nikkel standaarden
Ni in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.