VWR®, Nickel standard solution, 1000 mg/L Ni in dil. nitric acid AVS TITRINORM standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86696.260EA
47.1
EUR
86696.260
86696.180
VWR®, Nickel standard solution, 1000 mg/L Ni in dil. nitric acid AVS TITRINORM standard for AAS
Standaarden
Kationische standaarden
Nikkel standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%