VWR® ARISTAR®, Nickel Standard Solution, 10000 mg/L Ni in dil. Nitric Acid (from Ni), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455732GEA
316
EUR
455732G
455734Y
VWR® ARISTAR®, Nickel Standard Solution, 10000 mg/L Ni in dil. Nitric Acid (from Ni), Standard for ICP
Standaarden
Kationische standaarden
Nikkel standaarden
Ni in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.