VWR® AVS TITRINORM, Mercury Standard Solution, 1000 mg/l Hg in 10% Nitric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86682.260EA 32.97 EUR
86682.260 86682.180
VWR® AVS TITRINORM, Mercury Standard Solution, 1000 mg/l Hg in 10% Nitric Acid, Standard for AAS
Standaarden Kationische standaarden Kwik standaarden
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%