VWR® ARISTAR®, Mercury Standard Solution, 10000 mg/l Hg in dil. Nitric Acid (from Hg), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455672LEA 316 EUR
455672L 455674N
VWR® ARISTAR®, Mercury Standard Solution, 10000 mg/l Hg in dil. Nitric Acid (from Hg), Standard for ICP
Standaarden Kationische standaarden Kwik standaarden
Hg in HNO₃ 10%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.