VWR® ARISTAR®, Rhenium Standard Solution, 1000 mg/l Re in Water with Nitric Acid (max. 1%) (from Re), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455862PEA 377 EUR
455862P 455864R
VWR® ARISTAR®, Rhenium Standard Solution, 1000 mg/l Re in Water with Nitric Acid (max. 1%) (from Re), Standard for ICP
Standaarden Kationische standaarden Renium standaarden
Re in H₂O tr. HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with Certificate of Analysis.