VWR® ARISTAR®, Rhenium Standard Solution, 10000 mg/l Re in Water with Nitric Acid (max. 1%) (from Re), Standard for ICP
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VWR® ARISTAR®
455872REA 718 EUR
455872R
VWR® ARISTAR®, Rhenium Standard Solution, 10000 mg/l Re in Water with Nitric Acid (max. 1%) (from Re), Standard for ICP
Standaarden Kationische standaarden Renium standaarden
Re in H₂O tr. HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022