VWR® ARISTAR®, Rhodium Standard Solution, 1000 mg/l Rh in 10% Hydrochloric Acid (from RhCl3), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455882TEA 1050 EUR
455882T 455884V
VWR® ARISTAR®, Rhodium Standard Solution, 1000 mg/l Rh in 10% Hydrochloric Acid (from RhCl3), Standard for ICP
Standaarden Kationische standaarden Rodium standaarden
RhCl₃.3H₂O in HCl 10%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.